Bolts and clamp rings
Bolts and clamps made entirely of PTFE, which are mainly used for moon-shaped stirrer blades with double-sided groove.
- Temperature resistance: −200 to 250 °C
- Universal chemical resistance
Frequently Bought Together
Bolts and clamps made entirely of PTFE, which are mainly used for moon-shaped stirrer blades with double-sided groove.