Order Entry
Austria
Orders LinkContactUsLinkComponent
Pyridine ≥99%, TECHNICAL

Pyridine ≥99%, TECHNICAL

Glass bottle, 1 L
Catalog # 27194.295
Flammable
Irritant
Supplier:   VWR Chemicals
Pyridine ≥99%, TECHNICAL
Catalog # 27194.295
Supplier:  VWR Chemicals
CAS Number:  110-86-1
Pyridine ≥99%, TECHNICAL
Pyridine ≥99%, TECHNICAL

Glass bottle, 1 L

CAS Number:  110-86-1
Price
131,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.