4-Aminopyridine
Catalog # EHERC10222000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:94.12 g/mol
Merck Index:13,03964
Boiling Pt:273 °C (1013 hPa)
Melting Pt:157…160 °C
Density:1.25 g/cm³ (20 °C)
Flash Pt:156 °C
Storage Temperature:Ambient
EINECS:207-987-9
UN:2671
ADR:6.1,II
CAS Number:504-24-5
Formula:C₅H₆N₂
MDL Number:MFCD00006439