ContactUsLinkComponent
Adapters, receiver, BOLA
  BOHLB292-02
 :  Bohlender
undefined
Adapters, receiver, BOLA
  BOHLB292-02
 :  Bohlender
  • Cone size:
    29/32 NS
  • Socket size:
    19/26 NS
  • Pk:
    1
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
Ground joint cone and socket size NS 29, as well as socket size NS 19 for example for vacuum stopcock (see Cat. No. B 293-02). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.