Order Entry
Austria
ContactUsLinkComponent
Stirrer bearings, PTFE, BOLA
Stirrer bearings, PTFE, BOLA
  BOHLB288-02
 :  Bohlender
undefined
Stirrer bearings, PTFE, BOLA
  BOHLB288-02
 :  Bohlender

 

  • For:
    Shaft - Ø: 10 mm
  • Description:
    Stirrer bearing with gasket and nut
  • NS stopper:
    29/32
  • Pk:
    1

 

 

PTFE stirrer bearings with adjustable special gasket.

  • Chemical resistance
  • Temperature resistance: −20 to 250 °C

Integrated special nut is present for easy locking and unlocking of the ground joint.