Order Entry
Austria
ContactUsLinkComponent
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Irritant
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    ≥ 99.9 %
  • Identification (IR):
    Conforms
  • Acidity:
    ≤ 0.0002 meq/g
  • Alkalinity:
    ≤ 0.0002 meq/g
  • Peroxides (as H₂O₂):
    ≤ 50 ppm
  • Water:
    ≤ 0.02 %
  • Transmittance (218 nm):
    ≥ 30 %
  • Transmittance (230 nm):
    ≥ 40 %
  • Transmittance (240 nm):
    ≥ 60 %
  • Transmittance (250 nm):
    ≥ 70 %
  • Transmittance (260 nm):
    ≥ 80 %
  • Transmittance (280 nm):
    ≥ 96 %
  • Transmittance (300 nm):
    ≥ 98 %
  • Filtered through 0.2 μm:
    Confirmed

Specification Test Results

Frequently Bought Together