


Tetrahydrofuran
Catalog # MSPP412452
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356