Order Entry
Austria
ContactUsLinkComponent
Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®
Flammable
Health
Irritant
Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®
Catalog # 9439-12
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®
Catalog # 9439-12
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, septum-sealed
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-320 nm:
    ≤0.005
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.030
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:
    ≤0.15
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤212
  • Assay (by GC, corrected for water):
    ≥99.8 %
  • Color (APHA):
    ≤10
  • Peroxide (as H₂O₂) (at time of fill):
    ≤10 ppm
  • Residue after Evaporation:
    ≤2.0 ppm
  • Water (by KF, coulometric):
    ≤50 ppm

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356