Order Entry
Austria
ContactUsLinkComponent
Tetrahydrofuran ≥99.8% unstabilised, Biograde for DNA and peptides synthesis
Flammable
Health
Irritant
Tetrahydrofuran ≥99.8% unstabilised, Biograde for DNA and peptides synthesis
Catalog # 44505.K2
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99.8% unstabilised, Biograde for DNA and peptides synthesis
Catalog # 44505.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :72.11 g/mol
Merck Index:14,9211
Boiling Pt:66 °C
Melting Pt:–108 °C
Density:0.889
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356