


Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:14,9211
Boiling Pt:66 °C
Melting Pt:–108 °C
Density:0.889
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together