Order Entry
Austria
ContactUsLinkComponent
Contact plate, Sterilin®
Contact plate, Sterilin®
Catalog # 391-2031
Supplier:  Sterilin
undefined
Contact plate, Sterilin®
Catalog # 391-2031
Supplier:  Sterilin

Specifications

  • Ø:
    67 mm
  • Sterility:
    Aseptic
  • Version:
    Contact plate, 55 mm
  • Height:
    10 mm
  • Pk:
    300

Specifications

About this item

Ideal for use in routine hygiene monitoring of surfaces.

  • Concave profile of the dish raises the profile of the media when set giving better contact between the agar bed and surface under test
  • Aseptically manufactured under cleanroom conditions (class 7 ISO 14644) to exclude microbiological contamination
  • Triple vented to aid gaseous exchange
  • Numbered grid on the base facilitates colony counting
  • Deep skirted base aids stability when stacked

Dimensions shown are for outer Ø base and outer height (mm).

Certifications: Conforms to IES 1993 and IPF 1990 standards.