Order Entry
Austria
ContactUsLinkComponent
 
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
 
undefined
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters

 

  • Catalog No:
  • 662-1350
  • 662-2268
  • 662-2269
  • 662-2270
  • Model:
  • pH 110
  • pH 110 M
  • pH 110 M
  • pH 110 M
  • pH range:
  • −2,00...+16,00
  • −2,00...+16,00
  • −2,00...+16,00
  • −2,00...+16,00
  • pH resolution:
  • 0,01
  • 0,01
  • 0,01
  • 0,01
  • pH accuracy:
  • ±0,1% ±2 digits
  • ±0,01% of full scale or ±0,03 pH units
  • ±0,01% of full scale or ±0,03 pH units
  • ±0,01% of full scale or ±0,03 pH units
  • pH calibration:
  • 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
  • 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
  • 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
  • 2-point (First point must be a 6,86/7,00 buffer, the second either a 4,00/4,01 or 9,18/10,01)
  • Automatic buffer recognition:
  • US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
    NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
  • US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
    NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
  • US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
    NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
  • US (4,00/7,00/10,00 @ 25 °C, 4,00/7,02/10,06 @ 20 °C)
    NIST (4,01/6,86/9,18 @ 25 °C, 4,00/6,87/9,23 @ 20 °C)
  • mV range:
  • ±1999
  • ±1999
  • ±1999
  • ±1999
  • mV resolution:
  • 1
  • 1
  • 1
  • 1
  • mV accuracy:
  • ±0,1% ±1 digit
  • ±0,1% ±1 digit
  • ±0,1% ±1 digit
  • ±0,1% ±1 digit
  • Temperature range:
  • −10...+120 °C
  • −10...+120 °C
  • −10...+120 °C
  • −10...+120 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,3 °C ±2 digits
  • ±0,3 ±1 digit
  • ±0,3 ±1 digit
  • ±0,3 ±1 digit
  • Temperature compensation:
  • Automatic/manual (−10...+120 °C)
  • Automatic/manual (−10...+120 °C)
  • Automatic/manual (−10...+120 °C)
  • Automatic/manual (−10...+120 °C)
  • Power supply:
  • One 9 Volt battery
  • One 9 Volt battery
  • One 9 Volt battery
  • One 9 Volt battery
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Data logging capacity:
  • 50 data sets
  • 250 data sets
  • 250 data sets
  • 250 data sets
  • Outputs:
  • -
  • Micro USB
  • Micro USB
  • Micro USB
  • W×D×H:
  • 78×37×184 mm
  • 78×37×184 mm
  • 78×37×184 mm
  • 78×37×184 mm
  • Weight:
  • 0,272 kg
  • 0,272 kg
  • 0,272 kg
  • 0,272 kg

 

pH buffer capsule

 

Frequently Bought Together