Order Entry
Austria
Orders LinkContactUsLinkComponent
Micro pipette controller

Micro pipette controller

Micro pipettor, 1 ml
Catalog # MARI2960408
Supplier:   MARIENFELD
Micro pipette controller
Catalog # MARI2960408
Supplier:  MARIENFELD
Micro pipette controller
Micro pipette controller

Micro pipettor, 1 ml

Price
66,42 €
Log in for availability
1 * 1 ST
Product Details & Documents

This micro pipette filler is suitable for micro pipettes with circular mark and other small pipettes up to 1 ml capacity (for example: blood diluting pipettes).

  • Facilitates the sampling
  • Easy to use
  • Integrated ejection device reduces the risk of infection through contaminated pipettes
  • Autoclavable at 121 °C
Specifications
  • Capacity:
    1 ml
  • Description:
    Micro pipettor
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.