

Zinc oxide 99.0-100.5% (excluding ignited substances) USP, Macron Fine Chemicals™
:1314-13-2
:
:ZnO
:MFCD00011300
:81.39 g/mol
:13,10200
:1975 °C
:5.61 g/cm³ (25 °C)
:Ambient
:215-222-5
:3077
:9,III
:01-2119463881-32
:GHS09
:GHS07
- GMP Manufactured Product:-
- Meets U.S.P Requirements:-
- CAUTION: For Manufacturing, processing or repackaging:-
- Bulk Pharmaceutical Chemical:-
- USP - Assay (ZnO) (ignited basis):98.0 - 102.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Alkalinity:Passes Test
- USP - Loss on Ignition:≤1.0 %
- USP - Carbonate and Color of Solution:Passes Test
- USP - Arsenic (As):≤6 ppm
- USP - Lead (Pb):Passes Test
- USP - Iron and Other Heavy Metals:Passes Test
- Solubility in Dilute Sulfuric Acid:Passes Test
- Lead (Pb):≤0.0010 %
- Cadmium (Cd):≤0.0015 %
- Iron (Fe):≤0.0020 %
Frequently Bought Together