


Silver nitrate, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:7761-88-8
:
:AgNO₃
:MFCD00003414
:169.87 g/mol
:12,08661
:444 °C (1013 hPa)
:210 °C
:4.35 g/cm³ (20 °C)
:Ambient
:231-853-9
:1493
:5.1,II
:01-2119513705-43
:GHS03
:GHS05
:GHS09
- Assay:min. 99.9 %
- Clarity of Solution:passes test
- Lead (Pb):max. 0.001 %
- Free Acid:passes test
- Substances not Precipitated by HCl:max. 0.01 %
- Sulfate (SO4):max. 0.002 %
- Chloride (Cl):max. 5
- Copper (Cu):max. 2
- Iron (Fe):max. 2
Frequently Bought Together