
7-Isoquinolinol ≥97%
Catalog # L17067.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
CAS Number:7651-83-4
:
Formula:C₉H₇NO
MDL Number:MFCD00456131
Melting Pt:226…228 °C
:145.16 g/mol
:GHS07
Storage Temperature:Ambient
:14476-74-5