Order Entry
Austria
ContactUsLinkComponent
Ammonium nitrate ≥98.5%, GPR RECTAPUR®
Oxidizing
Ammonium nitrate ≥98.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Ammonium nitrate ≥98.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together