ContactUsLinkComponent
Silica gel, Pore size: ∼60 Å 70-230 mesh, high purity for column chromatography, Supelco®
Silica gel, Pore size: ∼60 Å 70-230 mesh, high purity for column chromatography, Supelco®
  60740-500G
 :  Merck
 :  
undefined
Silica gel, Pore size: ∼60 Å 70-230 mesh, high purity for column chromatography, Supelco®
  60740-500G
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
 :
 :60.08 g/mol
 :M-Clarity™ quality level = MQ100
 :Ambient
 :112926-00-8
 :7631-86-9
 :SiO₂
 :MFCD00011232