
Silica gel, Pore size: 60 Å 0.075-0.2 mm (70-200 mesh)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:1710 °C
Density:2.4
Storage Temperature:Ambient
EINECS:231-545-4
:GHS08
Related Information
Frequently Bought Together