Order Entry
Austria
ContactUsLinkComponent
Silica gel, Pore size: 60 Å 0.035-0.070 mm for chromatography
Silica gel, Pore size: 60 Å 0.035-0.070 mm for chromatography
Certificates
undefined
Silica gel, Pore size: 60 Å 0.035-0.070 mm for chromatography
Certificates

Specification Test Results

  • Appearance:
    White Powder
  • Particle size:
    0.035 to 0.070 mm
  • Specific gravity:
    0.46 to 0.55 kg/l (Typical)
  • pH:
    6.5 to 7.5 (5 % suspension in water) Typical
  • Residual water:
    ≤6 % (160°C, 15 min)

Specification Test Results

Related Information

Frequently Bought Together