Order Entry
Austria
ContactUsLinkComponent
Silica gel, Pore size: 100 Å 15-25 μm, high purity for column chromatography, Supelco®
Silica gel, Pore size: 100 Å 15-25 μm, high purity for column chromatography, Supelco®
Catalog # 60744-500G
Supplier:  Merck
CAS Number:  
undefined
Silica gel, Pore size: 100 Å 15-25 μm, high purity for column chromatography, Supelco®
Catalog # 60744-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :60.08 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:112926-00-8
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232