Order Entry
Austria
ContactUsLinkComponent
Silica gel, Pore size: 150 Å 0.075-0.250 mm for chromatography
Silica gel, Pore size: 150 Å 0.075-0.250 mm for chromatography
Certificates
undefined
Silica gel, Pore size: 150 Å 0.075-0.250 mm for chromatography

Specification Test Results

  • Appearance:
    White Granular powder
  • Particle size:
    ≤7.5 % > 60 Mesh
  • Specific surface area:
    275 to 375 m²/g (BET)
  • Pore volume:
    0.95 to 1.3 ml/g (N2)
  • Volatiles:
    ≤8.0 % (160°C)

Specification Test Results

Related Information

Frequently Bought Together