Order Entry
Austria
ContactUsLinkComponent
Orthoboric acid ≥99%, powder, GPR RECTAPUR®
Health
Orthoboric acid ≥99%, powder, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Orthoboric acid ≥99%, powder, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Non-volatile with methanol/HCl:
    Max. 0.2 %
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.025 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together