
Orthoboric acid ≥98%, crystallised, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
28041714.pdf spec_VWRC20177.290 spec_VWRC20177.368
Product specification
Frequently Bought Together