
Orthoboric acid, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 0501.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.0 - 100.5 %
- Appearance of solution:passes test
- Heavy Metals (as Pb):max. 15 ppm
- Identification:passes test
- Loss on Drying:max. 0.5 %
- Organic Matter:passes test
- pH:3.8 - 4.8
- Solubility in Alcohol:passes test
- Sulfates (as SO4):max. 450 ppm
Specification Test Results
About this item
:
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337