Order Entry
Austria
ContactUsLinkComponent
Diboron trioxide, Suprapur® for trace analysis, Supelco®
Diboron trioxide, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
Diboron trioxide, Suprapur® for trace analysis, Supelco®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    36 g/L (25 °C)
  • Melting Point:
    450 °C
  • Molar Mass:
    69.61 g/mol
  • Bulk Density:
    900 - 1100 kg/m³
  • Boiling Point:
    1860 °C
  • Density:
    2.46 g/cm³ (20 °C)
  • pH:
    4 (10 g/L, H₂O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together