0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Pipettes
Pipette Bulbs
Pipette teats
Pipette teats
391-8811
:
MARIENFELD
Pipette teats
391-8811
:
MARIENFELD
Colour:
Red
Description:
Rubber teats for dropping pipettes, with ∼0,8 ml suction volume
Environmentally Preferable:
Pk:
100
Rubber teats for dropping pipettes.