Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALG7294-100ML
Supplier:  Merck
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALG7294-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :95.53 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026