
Guanidinium thiocyanate 6 mol/l in aqueous solution for molecular biology, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
EINECS:209-812-1
:GHS06
Storage Temperature:Ambient
Frequently Bought Together