Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Guanidinium hydrochloride
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
SIAL50937-100ML
:
Merck
:
Guanidinium hydrochloride 8 mol/l in aqueous solution, Sigma-Aldrich®
SIAL50937-100ML
:
Merck
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
95.53 g/mol
:
200-002-3
:
GHS07
:
Ambient
:
50-01-1
:
NH₂C(=NH)NH₂·HCl
:
MFCD00013026