Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydrazinobenzoic acid hydrochloride
4-Hydrazinobenzoic acid hydrochloride, Sigma-Aldrich®
4-Hydrazinobenzoic acid hydrochloride, Sigma-Aldrich®
SIAL433829-25G
:
Merck
:
4-Hydrazinobenzoic acid hydrochloride, Sigma-Aldrich®
SIAL433829-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
188.61 g/mol
:
M-Clarity™ quality level = MQ100
:
246-330-0
:
24589-77-3
:
C₇H₉ClN₂O₂
:
MFCD00039073