ContactUsLinkComponent
Malachite green oxalate in water, crystalline for microscopy, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Malachite green oxalate in water, crystalline for microscopy, Sigma-Aldrich®
Certificates
undefined
Corrosive
Health
Irritant
Environment
Malachite green oxalate in water, crystalline for microscopy, Sigma-Aldrich®
Certificates
Specification Test Results
  • Solubility:
    0.01 g in 10 ml methanol
  • Loss on drying:
    ≤7.0%
  • Wavelength (1) UV:
    609 - 619 nm
  • Wavelength (2) UV:
    420 - 430 nm
  • Extinction (1%/1 cm) (1) (UV):
    ≥1450
Specification Test Results
Related Information
Certificates
Search by Lot Number
Frequently Bought Together