(S)-(+)-Ethyl mandelate, Sigma-Aldrich®
Catalog # SIAL309974-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:180.2 g/mol
CAS Number:13704-09-1
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00064249