(-)-N-Methylephedrine, Sigma-Aldrich®
Catalog # SIAL235210-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:179.26 g/mol
:M-Clarity™ quality level = MQ200
EINECS:209-022-7
CAS Number:552-79-4
Formula:C₁₁H₁₇NO
MDL Number:MFCD00004504