

Glyoxylic acid 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:298-12-4
:
Formula:C₂H₂O₃
MDL Number:MFCD00006958
:74.04 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:206-058-5
UN:3265
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:111 °C (1013 hPa)
- Vapor Pressure:10 hPa (25 °C)
- Flash Point:115 °C
- Refractive Index:1.4149 (20 °C, 589 nm)
- Density:1.34 g/cm3 (20 °C)
- pH:0.3 (H2O, 19.5 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.14084.0250_TDS_EN MERC8.14084.1000_TDS_EN
Product specification
msds_814084
SDS
Frequently Bought Together