Order Entry
Austria
ContactUsLinkComponent
Aluminium chloride for synthesis, Sigma-Aldrich®
Corrosive
Aluminium chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Aluminium chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    450 g/l (20 °C) (decomposition)
  • Melting Point:
    180 - 181 °C (sublimed)
  • Molar Mass:
    133.34 g/mol
  • Bulk Density:
    1200 kg/m3
  • Vapor Pressure:
    1 hPa (20 °C)
  • Density:
    2.44 g/cm3 (20 °C)
  • pH:
    2.4 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Frequently Bought Together