4-Hydrazinobenzoic acid hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24589-77-3
:
Formula:C₇H₉ClN₂O₂
MDL Number:MFCD00039073
:188.61 g/mol
Melting Pt:252…254 °C (dec.)
EINECS:246-330-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together