Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution standardised solution 1.0 N

Ammonia solution standardised solution 1.0 N

Glass bottle, 1 L
Catalog # 35614.K2
Corrosive
Ammonia solution standardised solution 1.0 N
Catalog # 35614.K2
CAS Number:  1336-21-6
Ammonia solution standardised solution 1.0 N
Ammonia solution standardised solution 1.0 N

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
77,20 €
Log in for availability
1 * 1 L
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Appearance: Colourless Odour: Ammonia-like MDL No.: MFCD00066650

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.