
Platinum 5% on activated carbon hydrogenation catalyst
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-06-4
:
Formula:Pt
MDL Number:MFCD00011179
:195.08 g/mol
Storage Temperature:Ambient
EINECS:231-116-1
UN:1325
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Black Powder
- Assay:5 % (Pt)
- Additional info:typical properties:
Specification Test Results
Frequently Bought Together