Orders LinkContactUsLinkComponent
Preparation table for slides
Preparation table for slides
Tables de préparation numérotée, White, Cardboard, 340×205×5 mm
Catalog # 630-2771
Supplier:   MARIENFELD
Preparation table for slides
Catalog # 630-2771
Supplier:  MARIENFELD
Preparation table for slides
Preparation table for slides
Tables de préparation numérotée, White, Cardboard, 340×205×5 mm
Price
148,55 €
Log in for availability
1 * 20 ST
Product Details & Documents
Preparation table for 20 microscopy slides. Thanks to its recessed grips the slides can be easily taken out.
Specifications
  • Colour:
    White
  • Description:
    Tables de préparation numérotée
  • Material:
    Cardboard
  • W×D×H:
    340×205×5 mm