Orders LinkContactUsLinkComponent
Dishes
Dishes
Supplier:   KARTELL
Dishes
Supplier:  KARTELL
Certificates
Dishes
2 Options Available Below
Product Details & Documents
Made of PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Good mechanical resistance
Dishes are suitable for foodstuff. It can withstand temperatures continuously up to 250 °C and for short intervals up to 300 °C.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 25 ml
    25 mm
    42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,10 €
    47,10 €
    1 * 1 ST
       47,10 €
  • 100 ml
    28 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €