Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Sulphuric acid 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Supplier:  Merck
Certificates
Sulphuric acid 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Melting Point:
    -20 °C
  • Molar Mass:
    98.08 g/mol
  • Boiling Point:
    335 °C
  • Vapor Pressure:
    0.0001 hPa (20 °C)
  • Density:
    1.84 g/cm3 (20 °C)
  • pH:
    0.3 (49 g/l, H2O, 25 °C)

Specification Test Results

Frequently Bought Together