
Sulphuric acid 95.0-98.0% (by alkalimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, JPE, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:335 °C (1013 hPa)
Melting Pt:–20 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
Specification Test Results
- Assay (Alkalimetric):95.0 - 98.0%
- Identity:Passes Test
- Appearance of Solution:Passes Test
- Chloride (Cl):≤0.0002%
- Nitrate (NO₃):≤0.00010%
- Nitrate (NO₃) (Ph. Eur.):Passes Test
- Heavy Metals (as Pb):≤0.0005%
- As (Arsenic):≤0.000010%
- Fe (Iron):≤0.0002%
- NH₄ (Ammonium):≤0.0005%
- Pb (Lead):≤0.0005%
- V (Vanadium):≤0.0002
- Residual Solvents (ICH Q3C):Excluded by Production Process
- Substances Reducing Potassium Permanganate (as SO₂):≤0.0010%
- Residue on Ignition:≤0.0050%
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.00713.1000_TDS_EN ProductPage_100713
Product specification
msds_100713
SDS
Frequently Bought Together