Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 95-97%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Sulphuric acid 95-97%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Sulphuric acid 95-97%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    95 - 97 %
  • Ammonium (NH4):
    max. 1 ppm
  • Color (APHA):
    max. 10
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Nitrate (NO3):
    max. 1 ppm
  • Residue after Ignition:
    max. 5 ppm
  • Substances Reducing KMnO4(as SO2):
    max. 5 ppm
  • Silver (Ag):
    max. 0.01
  • Aluminium (Al):
    max. 0.05
  • Arsenic (As):
    max. 0.05
  • Barium (Ba):
    max. 0.02
  • Beryllium (Be):
    max. 0.01
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.02
  • Chloride (Cl):
    max. 0.5
  • Cobalt (Co):
    max. 0.01
  • Chromium (Cr):
    max. 0.1
  • Copper (Cu):
    max. 0.01
  • Iron (Fe):
    max. 0.1
  • Germanium (Ge):
    max. 0.05
  • Potassium (K):
    max. 0.1
  • Lithium (Li):
    max. 0.01
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.01
  • Molybdenum (Mo):
    max. 0.02
  • Sodium (Na):
    max. 0.5
  • Nickel (Ni):
    max. 0.05
  • Lead (Pb):
    max. 0.05
  • Strontium (Sr):
    max. 0.01
  • Titanium (Ti):
    max. 0.1
  • Thallium (Tl):
    max. 0.05
  • Vanadium (V):
    max. 0.01
  • Zinc (Zn):
    max. 0.05
  • Zirconium (Zr):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together