Sodium sulphate, anhydrous, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, extra pure, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7757-82-6
:
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
Related Information
Frequently Bought Together