Order Entry
Austria
ContactUsLinkComponent
Sodium sulphate, anhydrous, granulated, EMSURE® for organic trace analysis, Supelco®
Sodium sulphate, anhydrous, granulated, EMSURE® for organic trace analysis, Supelco®
Catalog # 1.06639.0500
Supplier:  Merck
CAS Number:  
undefined
Sodium sulphate, anhydrous, granulated, EMSURE® for organic trace analysis, Supelco®
Catalog # 1.06639.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    small crystals
  • Assay (acidimetric):
    ≥ 99.0 %
  • Fe (Iron):
    ≤ 0.001 %
  • Loss on drying (130 °C):
    ≤ 0.5 %

Specification Test Results

About this item

 :
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504