Order Entry
Austria
ContactUsLinkComponent
 
Sodium sulphate, anhydrous 99%, extra pure
 
undefined
Sodium sulphate, anhydrous 99%, extra pure
 

 

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥98.5 %
  • Water:
    ≤0.1 %
  • Sodium chloride (NaCl):
    ≤3000 ppm
  • Iron (Fe):
    ≤10 ppm
  • Zinc (Zn):
    ≤200 ppm

 

Frequently Bought Together