Sodium dithionite ∼ 85% tech.
: Sodium hydrosulphite
:7775-14-6
:
:Na₂S₂O₄
:MFCD00011640
:174.11 g/mol
:13,08700
:>300 °C (dec.)
:2,19 g/cm³ (20 °C)
:101 °C
:Ambient
:231-890-0
:1384
:4.2,II
:GHS07
:GHS02
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Titration with NaOH:≥85 %
- Titration Argentometric:≥96.0 to ≤104.0 %
- HPLC:≥96.0 %
Frequently Bought Together