Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous, EMSURE® (0.2 - 1 mm) for analysis, Supelco®
Disodium hydrogen phosphate, anhydrous, EMSURE® (0.2 - 1 mm) for analysis, Supelco®
Supplier:  Merck
Certificates
Disodium hydrogen phosphate, anhydrous, EMSURE® (0.2 - 1 mm) for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    77 g/l (20 °C)
  • Melting Point:
    250 °C (decomposition)
  • Molar Mass:
    141.96 g/mol
  • Bulk Density:
    880 kg/m3
  • pH:
    8.7 - 9.3 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together