Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Disodium hydrogen phosphate, anhydrous ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Disodium hydrogen phosphate, anhydrous ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • pH (25°C; 5 %):
    8.7 - 9.3
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Loss on drying (105°C):
    Max. 0.2 %
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Cu (Copper):
    Max. 3 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 100 ppm
  • Pb (Lead):
    Max. 10 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together